ENA295068996 Enamine

6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline-2-sulfonamide

  • MDL number MFCD11205212
  • Cas Number : 1094222-85-1
  • Molecular Weight : 272.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-62661

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62661
Availability details for
Quantity
  • Availability
  • Add to Cart