ENA295069001 Enamine

6-fluoro-4-(trifluoromethyl)quinoline-3-carboxylic acid

  • MDL number MFCD09040799
  • Cas Number : 596845-53-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62663

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62663
Availability details for
Quantity
  • Availability
  • Add to Cart