ENA295069011 Enamine

6-chloro-8-methyl-3,4-dihydro-2H-1-benzopyran-4-one

  • MDL number MFCD11207527
  • Cas Number : 76301-90-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62673

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62673
Availability details for
Quantity
  • Availability
  • Add to Cart