ENA295069014 Enamine

2-(1-phenyl-1H-pyrazol-3-yl)ethan-1-amine

  • MDL number MFCD02853702
  • Cas Number : 1240526-44-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-62675

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62675
Availability details for
Quantity
  • Availability
  • Add to Cart