ENA295069019 Enamine

2-amino-2-methyl-N-[3-(pyrrolidin-1-yl)phenyl]pentanamide

  • MDL number MFCD16547570
  • Cas Number : 1179625-10-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62679

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62679
Availability details for
Quantity
  • Availability
  • Add to Cart