ENA295069020 Enamine

1-benzyl-2-(pyrrolidin-2-yl)pyrrolidine

  • MDL number MFCD16547571
  • Cas Number : 1240527-62-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62681

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62681
Availability details for
Quantity
  • Availability
  • Add to Cart