ENA295069033 Enamine

5-oxo-5-[4-(1,3-thiazol-2-yl)piperazin-1-yl]pentanoic acid

  • MDL number MFCD10694889
  • Cas Number : 1152583-28-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62724

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62724
Availability details for
Quantity
  • Availability
  • Add to Cart