ENA295069034 Enamine

6-[ethyl(propan-2-yl)amino]pyridine-3-carboxylic acid

  • MDL number MFCD11624749
  • Cas Number : 1094798-46-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62725

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62725
Availability details for
Quantity
  • Availability
  • Add to Cart