ENA295069037 Enamine

3-(cycloheptylsulfamoyl)thiophene-2-carboxylic acid

  • MDL number MFCD09455214
  • Cas Number : 944894-50-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62729

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62729
Availability details for
Quantity
  • Availability
  • Add to Cart