ENA295069038 Enamine

3-[(2-hydroxy-2-phenylethyl)sulfamoyl]thiophene-2-carboxylic acid

  • MDL number MFCD13714544
  • Cas Number : 1155165-61-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62730

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62730
Availability details for
Quantity
  • Availability
  • Add to Cart