ENA295069039 Enamine

4-(2-cyclopentylacetamido)cyclohexane-1-carboxylic acid

  • MDL number MFCD13167836
  • Cas Number : 1184211-85-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62731

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62731
Availability details for
Quantity
  • Availability
  • Add to Cart