ENA295069044 Enamine

1-(naphthalen-2-yl)propan-2-amine

  • MDL number MFCD08452682
  • Cas Number : 18085-03-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62738

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62738
Availability details for
Quantity
  • Availability
  • Add to Cart