ENA295069045 Enamine

N-(2-aminoethyl)-2-cyclopentylacetamide

  • MDL number MFCD09728466
  • Cas Number : 953720-71-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62740

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62740
Availability details for
Quantity
  • Availability
  • Add to Cart