ENA295069058 Enamine

3-(cyclohexylformamido)propanoic acid

  • MDL number MFCD09050507
  • Cas Number : 914773-52-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62802

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62802
Availability details for
Quantity
  • Availability
  • Add to Cart