ENA295069059 Enamine

3-(chloromethyl)-2H-1lambda6,2,4-benzothiadiazine-1,1-dione

  • MDL number MFCD03018284
  • Cas Number : 37162-65-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 225-226
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-62803

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62803
Availability details for
Quantity
  • Availability
  • Add to Cart