ENA295069073 Enamine

2,2,2-trifluoroethyl N-[3-(furan-2-amido)phenyl]carbamate

  • MDL number MFCD16547509
  • Cas Number : 1223299-00-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56737

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56737
Availability details for
Quantity
  • Availability
  • Add to Cart