ENA295069077 Enamine

3-[(pyridin-3-yl)sulfamoyl]thiophene-2-carboxylic acid

  • MDL number MFCD09455227
  • Cas Number : 944894-10-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62818

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62818
Availability details for
Quantity
  • Availability
  • Add to Cart