ENA295069078 Enamine

[5-(3-chlorophenyl)-1,3-oxazol-2-yl]methanol

  • MDL number MFCD12137987
  • Cas Number : 1155531-69-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62819

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62819
Availability details for
Quantity
  • Availability
  • Add to Cart