ENA295069080 Enamine

N,N-dimethyl-1,3-thiazolidine-4-carboxamide

  • MDL number MFCD09945714
  • Cas Number : 1103879-74-8
  • Molecular Weight : 160.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62821

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62821
Availability details for
Quantity
  • Availability
  • Add to Cart