ENA295069082 Enamine

2,2,2-trifluoroethyl N-[4-(2,6-dimethylmorpholin-4-yl)-3-fluorophenyl]carbamate

  • MDL number MFCD13604719
  • Cas Number : 1208668-99-8
  • Molecular Weight : 350.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56816

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56816
Availability details for
Quantity
  • Availability
  • Add to Cart