ENA295069084 Enamine

4-{[3-(pyrrolidin-1-yl)phenyl]carbamoyl}butanoic acid

  • MDL number MFCD16547579
  • Cas Number : 1178270-35-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62825

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62825
Availability details for
Quantity
  • Availability
  • Add to Cart