ENA296274781 Enamine

4-(piperidin-1-yl)butanimidamide dihydrochloride

  • MDL number MFCD16622022
  • Cas Number : 1240526-76-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Product Link https://www.enaminestore.com/catalog/EN300-59113

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59113
Availability details for
Quantity
  • Availability
  • Add to Cart