ENA296274784 Enamine

1,4-difluoro-2-trifluoromethanesulfonylbenzene

  • MDL number MFCD16622025
  • Cas Number : 1240526-79-7
  • Molecular Weight : 246.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60821

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60821
Availability details for
Quantity
  • Availability
  • Add to Cart