ENA296274794 Enamine

3-ethyl-4-methyl-4H-1,2,4-triazole

  • MDL number MFCD16622035
  • Cas Number : 1034197-72-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64084

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64084
Availability details for
Quantity
  • Availability
  • Add to Cart