ENA296274795 Enamine

4-methyl-3-(propan-2-yl)-4H-1,2,4-triazole

  • MDL number MFCD16622036
  • Cas Number : 1240527-41-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64087

Safety Information

msds https://www.enaminestore.com/catalog/EN300-64087
Availability details for
Quantity
  • Availability
  • Add to Cart