ENA296274798 Enamine

2-(chloromethyl)-4-ethylmorpholine

  • MDL number MFCD16622040
  • Cas Number : 1240529-25-2
  • Molecular Weight : 163.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64154

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64154
Availability details for
Quantity
  • Availability
  • Add to Cart