ENA296274803 Enamine

3-amino-4-(pyrrolidin-1-yl)-1lambda6-thiolane-1,1-dione dihydrochloride

  • MDL number MFCD16622045
  • Cas Number : 1240528-69-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64441

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64441
Availability details for
Quantity
  • Availability
  • Add to Cart