ENA296274804 Enamine

(3-phenyl-1,2,4-oxadiazol-5-yl)methanediol

  • MDL number MFCD16622046
  • Cas Number : 73217-59-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64485

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64485
Availability details for
Quantity
  • Availability
  • Add to Cart