ENA296274805 Enamine

(4-methylquinazolin-2-yl)methanamine

  • MDL number MFCD16622048
  • Cas Number : 1024000-61-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-64549

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64549
Availability details for
Quantity
  • Availability
  • Add to Cart