ENA296274813 Enamine

(1-cyclopropylethyl)({[1,2,4]triazolo[4,3-a]pyridin-3-ylmethyl})amine hydrochloride

  • MDL number MFCD16622057
  • Cas Number : 1240527-26-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64791

Safety Information

msds https://www.enaminestore.com/catalog/EN300-64791
Availability details for
Quantity
  • Availability
  • Add to Cart