ENA296274814 Enamine

3-chloropropane-1-sulfonyl fluoride

  • MDL number MFCD16622058
  • Cas Number : 3821-80-5
  • Molecular Weight : 160.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64799

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64799
Availability details for
Quantity
  • Availability
  • Add to Cart