ENA296274815 Enamine

2-(1,3-dihydro-2-benzofuran-5-sulfonamido)acetic acid

  • MDL number MFCD16622059
  • Cas Number : 1240527-08-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64806

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64806
Availability details for
Quantity
  • Availability
  • Add to Cart