ENA296274820 Enamine

1-(4-fluorophenyl)-1-[3-(trifluoromethyl)phenyl]methanamine hydrochloride

  • MDL number MFCD16622065
  • Cas Number : 1240528-82-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64913

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64913
Availability details for
Quantity
  • Availability
  • Add to Cart