ENA296274822 Enamine

2-(oxolan-2-yl)ethan-1-amine hydrochloride

  • MDL number MFCD16622067
  • Cas Number : 1240527-33-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64931

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64931
Availability details for
Quantity
  • Availability
  • Add to Cart