ENA296274825 Enamine

(3,6-dichloropyridin-2-yl)methanol

  • MDL number MFCD13185536
  • Cas Number : 58804-10-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64991

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64991
Availability details for
Quantity
  • Availability
  • Add to Cart