ENA296274826 Enamine

4-amino-1,1,1-trifluoro-2-phenylbutan-2-ol

  • MDL number MFCD16622070
  • Cas Number : 1240528-87-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110...112
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64999

Safety Information

msds https://www.enaminestore.com/catalog/EN300-64999
Availability details for
Quantity
  • Availability
  • Add to Cart