ENA296274827 Enamine

2-[1-(3-fluorophenyl)-1H-pyrazol-3-yl]ethan-1-amine

  • MDL number MFCD16622071
  • Cas Number : 1240527-51-8
  • Molecular Weight : 205.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65028

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65028
Availability details for
Quantity
  • Availability
  • Add to Cart