ENA296274829 Enamine

5-chloro-6-[(3,4-difluorophenyl)amino]pyridine-3-carbonitrile

  • MDL number MFCD16622073
  • Cas Number : 1240526-18-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65080

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65080
Availability details for
Quantity
  • Availability
  • Add to Cart