ENA296274839 Enamine

3-(4-chlorophenyl)pentan-3-amine hydrochloride

  • MDL number MFCD16622085
  • Cas Number : 667864-91-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-65133

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65133
Availability details for
Quantity
  • Availability
  • Add to Cart