ENA296274844 Enamine

N-(3-chloro-4-cyanophenyl)(propan-2-yloxy)carbothioamide

  • MDL number MFCD16622090
  • Cas Number : 1240527-46-1
  • Molecular Weight : 254.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65163

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65163
Availability details for
Quantity
  • Availability
  • Add to Cart