ENA296274845 Enamine

4-(4-methyl-1,3-thiazol-2-yl)piperidine dihydrochloride

  • MDL number MFCD16622091
  • Cas Number : 1240528-17-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65167

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65167
Availability details for
Quantity
  • Availability
  • Add to Cart