ENA296274848 Enamine

methyl 3-cyclopropyl-7-oxo-6H,7H-[1,2]oxazolo[3,4-d]pyridazine-4-carboxylate

  • MDL number MFCD16622095
  • Cas Number : 1240527-42-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65182

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65182
Availability details for
Quantity
  • Availability
  • Add to Cart