ENA296274850 Enamine

[(1,3-dioxaindan-4-yl)methyl](ethyl)amine hydrochloride

  • MDL number MFCD16622097
  • Cas Number : 1240526-40-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65192

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65192
Availability details for
Quantity
  • Availability
  • Add to Cart