ENA296274854 Enamine

2-methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-6-sulfonamide

  • MDL number MFCD16622101
  • Cas Number : 1240527-49-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65209

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65209
Availability details for
Quantity
  • Availability
  • Add to Cart