ENA296274856 Enamine

2-amino-N-[4-(2H-1,2,3-triazol-2-yl)phenyl]acetamide hydrochloride

  • MDL number MFCD16622103
  • Cas Number : 1240527-83-6
  • Molecular Weight : 253.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65231

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65231
Availability details for
Quantity
  • Availability
  • Add to Cart