ENA296274857 Enamine

[(4-methoxy-3-nitrophenyl)methyl](methyl)amine hydrochloride

  • MDL number MFCD16622105
  • Cas Number : 346705-17-7
  • Molecular Weight : 232.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65235

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65235
Availability details for
Quantity
  • Availability
  • Add to Cart