ENA296274865 Enamine

1-[3-(aminomethyl)phenyl]-3-cyclopropylurea hydrochloride

  • MDL number MFCD16622114
  • Cas Number : 1240527-87-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65300

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65300
Availability details for
Quantity
  • Availability
  • Add to Cart