ENA296274866 Enamine

1-(3-aminophenyl)-5-methylimidazolidin-2-one

  • MDL number MFCD16622115
  • Cas Number : 1240528-94-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-65301

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65301
Availability details for
Quantity
  • Availability
  • Add to Cart