ENA296274869 Enamine

1-[(3-phenyl-1,2-oxazol-5-yl)methyl]piperazine dihydrochloride

  • MDL number MFCD16622118
  • Cas Number : 1008774-63-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65314

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65314
Availability details for
Quantity
  • Availability
  • Add to Cart