ENA296274872 Enamine

1,3-dimethyl-5-(pyridin-3-yloxy)-1H-pyrazol-4-amine dihydrochloride

  • MDL number MFCD16622121
  • Cas Number : 1240528-97-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65328

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65328
Availability details for
Quantity
  • Availability
  • Add to Cart